COMH4211AB1F Combi-Blocks, Inc.

1-[5-Fluoro-2-(2-methylpropoxy)phenyl]ethan-1-amine

  • MDL number MFCD11137229
  • Cas Number : 1019554-62-1
  • Molecular Weight : 211.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1172
Availability details for
Quantity
  • Availability
  • Add to Cart