COMH4211AB20 Combi-Blocks, Inc.

(R)-(1-Carbamoyl-piperidin-3-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD31583365
  • Cas Number : 2270918-23-3
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1174

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1174
Availability details for
Quantity
  • Availability
  • Add to Cart