COMH4211AB21 Combi-Blocks, Inc.

(10-Amino-decyl)-carbamic acid benzyl ester

  • MDL number MFCD02094516
  • Cas Number : 62196-18-3
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1180

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1180
Availability details for
Quantity
  • Availability
  • Add to Cart