COMH4211AB23 Combi-Blocks, Inc.

3-Bromo-2-ethoxyaniline

  • MDL number MFCD24118719
  • Cas Number : 1512352-61-2
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1182
Availability details for
Quantity
  • Availability
  • Add to Cart