COMH4211AB25 Combi-Blocks, Inc.

Trans ethanesulfonic acid (4-amino-cyclohexyl)-amide

  • MDL number MFCD22403090
  • Cas Number : 787549-14-8
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1190
Availability details for
Quantity
  • Availability
  • Add to Cart