COMH4211AB26 Combi-Blocks, Inc.

(4-Bromo-2-methyl-benzyl)-cyclopropyl-amine

  • MDL number MFCD12096723
  • Cas Number : 345966-00-9
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1192

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1192
Availability details for
Quantity
  • Availability
  • Add to Cart