COMH4211AB29 Combi-Blocks, Inc.

4-(4-Hydroxymethyl-phenoxymethyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31618932
  • Cas Number : 1809267-01-3
  • Molecular Weight : 321.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1198

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1198
Availability details for
Quantity
  • Availability
  • Add to Cart