COMH4211AB2A Combi-Blocks, Inc.

2-Bromo-4-ethenyl-1-fluorobenzene

  • MDL number MFCD20384567
  • Cas Number : 701914-09-2
  • Molecular Weight : 201.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1201
Availability details for
Quantity
  • Availability
  • Add to Cart