COMH4211AB2B Combi-Blocks, Inc.

1-(2-Chloro-3-(trifluoromethyl)benzyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazole

  • MDL number MFCD28718186
  • Cas Number : 1604036-98-7
  • Molecular Weight : 386.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1205

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1205
Availability details for
Quantity
  • Availability
  • Add to Cart