COMH4211AB2C Combi-Blocks, Inc.

5-(2-Hydroxy-ethylcarbamoyl)-pentanoic acid tert-butyl ester

  • MDL number MFCD31381620
  • Cas Number : 2204562-77-4
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1209

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1209
Availability details for
Quantity
  • Availability
  • Add to Cart