COMH4211AB2E Combi-Blocks, Inc.

1-(4-Fluoro-2-isobutoxyphenyl)ethanone

  • MDL number MFCD14635233
  • Cas Number : 1341880-01-0
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1216

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1216
Availability details for
Quantity
  • Availability
  • Add to Cart