COMH4211AB2F Combi-Blocks, Inc.

2-Methyl-3-(1h-pyrrol-2-yl)-propionic acid

  • MDL number MFCD30317059
  • Cas Number : 1520882-76-1
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1225

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1225
Availability details for
Quantity
  • Availability
  • Add to Cart