COMH4211AB30 Combi-Blocks, Inc.

4-(2,4-Difluorophenoxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

  • MDL number MFCD30721499
  • Cas Number : 1446236-84-5
  • Molecular Weight : 347.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1226
Availability details for
Quantity
  • Availability
  • Add to Cart