COMH4211AB32 Combi-Blocks, Inc.

3-(3-Cyano-4-fluorophenyl)propanoic acid

  • MDL number MFCD15527352
  • Cas Number : 412961-44-5
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1233

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1233
Availability details for
Quantity
  • Availability
  • Add to Cart