COMH4211AB33 Combi-Blocks, Inc.

5-Ethynyl-2,2-difluoro-benzo[1,3]dioxole

  • MDL number MFCD29079311
  • Cas Number : 1551182-90-1
  • Molecular Weight : 182.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1234

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1234
Availability details for
Quantity
  • Availability
  • Add to Cart