COMH4211AB34 Combi-Blocks, Inc.

4-Chloro-2-(pyrrolidin-1-ylmethyl)aniline

  • MDL number MFCD21957646
  • Cas Number : 859155-77-4
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1236
Availability details for
Quantity
  • Availability
  • Add to Cart