COMH4211AB35 Combi-Blocks, Inc.

6-Chloro-2-iodo-3-(2-(thiophen-3-yl)ethoxy)pyridine

  • MDL number MFCD27942965
  • Cas Number : 1282528-85-1
  • Molecular Weight : 365.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1239
Availability details for
Quantity
  • Availability
  • Add to Cart