COMH4211AB36 Combi-Blocks, Inc.

Carbonic acid isobutyl ester 4-nitro-phenyl ester

  • MDL number MFCD00775694
  • Cas Number : 112240-73-0
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1242

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1242
Availability details for
Quantity
  • Availability
  • Add to Cart