COMH4211AB37 Combi-Blocks, Inc.

3-(3-Fluorophenyl)-2-methylpropan-1-ol

  • MDL number MFCD20385974
  • Cas Number : 1339642-82-8
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1243

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1243
Availability details for
Quantity
  • Availability
  • Add to Cart