COMH4211AB39 Combi-Blocks, Inc.

5-(4-Methylpentyl)-1h-pyrazole-3-carboxylic acid ethyl ester

  • MDL number MFCD26385984
  • Cas Number : 856256-68-3
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1246
Availability details for
Quantity
  • Availability
  • Add to Cart