COMH4211AB3A Combi-Blocks, Inc.

3-Fluoro-4-(2,2,2-trifluoro-ethoxy)-benzylamine hydrochloride

  • MDL number MFCD29079286
  • Cas Number : 2365419-05-0
  • Molecular Weight : 259.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1247
Availability details for
Quantity
  • Availability
  • Add to Cart