COMH4211AB3C Combi-Blocks, Inc.

N-Ethyl-4-hydroxy-n,2-dimethylbenzamide

  • MDL number MFCD31613836
  • Cas Number : 1699998-81-6
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1249
Availability details for
Quantity
  • Availability
  • Add to Cart