COMH4211AB3D Combi-Blocks, Inc.

4-Ethyl-4-hydroxy-7,8-dihydro-1h-pyrano[3,4-f]indolizine-3,6,10(4h)-trione

  • MDL number MFCD22494933
  • Cas Number : 102978-40-5
  • Molecular Weight : 263.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1252

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1252
Availability details for
Quantity
  • Availability
  • Add to Cart