COMH4211AB3E Combi-Blocks, Inc.

5-Fluoro-3-n-propoxybenzyl alcohol

  • MDL number MFCD22372934
  • Cas Number : 1443344-90-8
  • Molecular Weight : 184.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1254

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1254
Availability details for
Quantity
  • Availability
  • Add to Cart