COMH4211AB40 Combi-Blocks, Inc.

5-Methyl-n-(piperidin-4-yl)furan-2-carboxamide

  • MDL number MFCD11170286
  • Cas Number : 1042652-03-8
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-N-(4-piperidinyl)-2-furamide
Inchi Code 1S/C11H16N2O2/c1-8-2-3-10(15-8)11(14)13-9-4-6-12-7-5-9/h2-3,9,12H,4-7H2,1H3,(H,13,14)
Inchi Key ZXMUDKONXBNYRB-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1257

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1257
Availability details for
Quantity
  • Availability
  • Add to Cart