COMH4211AB41 Combi-Blocks, Inc.

4-Hydroxy-pyrazole-1-sulfonic acid dimethylamide

  • MDL number MFCD31583370
  • Cas Number : 2270908-00-2
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1261
Availability details for
Quantity
  • Availability
  • Add to Cart