COMH4211AB42 Combi-Blocks, Inc.

N-tert-Butyl-4-chloro-2-methylbenzamide

  • MDL number MFCD31579491
  • Cas Number : 923977-54-2
  • Molecular Weight : 225.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1262
Availability details for
Quantity
  • Availability
  • Add to Cart