COMH4211AB43 Combi-Blocks, Inc.

(2S)-2-Methyl-1-(propane-2-sulfonyl)piperazine

  • MDL number MFCD24301031
  • Cas Number : 1567989-91-6
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1264
Availability details for
Quantity
  • Availability
  • Add to Cart