COMH4211AB45 Combi-Blocks, Inc.

3-Amino-5-cyclopropylmethoxy-n-methyl-benzamide

  • MDL number MFCD31614087
  • Cas Number : 2270907-55-4
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1266
Availability details for
Quantity
  • Availability
  • Add to Cart