COMH4211AB47 Combi-Blocks, Inc.

5-(4-Chloro-2-fluorophenyl)pyrimidin-2-amine

  • MDL number MFCD11877134
  • Cas Number : 1111111-97-7
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1269
Availability details for
Quantity
  • Availability
  • Add to Cart