COMH4211AB48 Combi-Blocks, Inc.

1-(5-Chloro-2-cyclopropylmethoxy-phenyl)-ethylamine hydrochloride

  • MDL number MFCD28963466
  • Molecular Weight : 262.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1270
Availability details for
Quantity
  • Availability
  • Add to Cart