COMH4211AB49 Combi-Blocks, Inc.

tert-Butyl (2-(2-(2-(2-nitrophenylsulfonamido)ethoxy)ethoxy)ethyl)carbamate

  • MDL number MFCD31579560
  • Cas Number : 943601-78-3
  • Molecular Weight : 433.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1271

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1271
Availability details for
Quantity
  • Availability
  • Add to Cart