COMH4211AB4A Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-(4-(4-((tetrahydro-2h-pyran-2-yl)oxy)butoxy)-phenyl)-1,3,2-dioxaborolane

  • MDL number MFCD31579499
  • Cas Number : 1210827-89-6
  • Molecular Weight : 376.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1274

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1274
Availability details for
Quantity
  • Availability
  • Add to Cart