COMH4211AB5E Combi-Blocks, Inc.

4'-Chloro-2'-fluoro-4-(trifluoromethoxy)biphenyl-3-carbaldehyde

  • MDL number MFCD31583176
  • Cas Number : 923281-75-8
  • Molecular Weight : 318.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1278
Availability details for
Quantity
  • Availability
  • Add to Cart