COMH4211AB60 Combi-Blocks, Inc.

2-Methyl-5-(prop-2-en-1-yloxy)aniline

  • MDL number MFCD11934502
  • Cas Number : 1175836-96-0
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1280

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1280
Availability details for
Quantity
  • Availability
  • Add to Cart