COMH4211AB64 Combi-Blocks, Inc.

Methyl 4-(4-cyanophenyl)-1h-pyrrole-2-carboxylate

  • MDL number MFCD31619002
  • Cas Number : 2102858-87-5
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1282
Availability details for
Quantity
  • Availability
  • Add to Cart