COMH4211AB66 Combi-Blocks, Inc.

4-(2-Fluoro-4-isobutoxy-phenyl)-1,2,3,6-tetrahydro-pyridine hydrochloride

  • MDL number MFCD31583387
  • Cas Number : 2270906-31-3
  • Molecular Weight : 285.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1284

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1284
Availability details for
Quantity
  • Availability
  • Add to Cart