COMH4211AB68 Combi-Blocks, Inc.

2-Chloro-6-(cyclopentylsulfanyl)pyridine

  • MDL number MFCD18368431
  • Cas Number : 122733-95-3
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1292
Availability details for
Quantity
  • Availability
  • Add to Cart