COMH4211AB6C Combi-Blocks, Inc.

Carbamic acid, [2-oxo-2-(2-propynylamino)ethoxy]-, 1,1-dimethylethyl ester

  • MDL number MFCD24474433
  • Cas Number : 203435-40-9
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1298

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1298
Availability details for
Quantity
  • Availability
  • Add to Cart