COMH4211AB6E Combi-Blocks, Inc.

Icosanoic acid iodomethyl ester

  • MDL number MFCD31583272
  • Cas Number : 1314041-19-4
  • Molecular Weight : 452.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1299
Availability details for
Quantity
  • Availability
  • Add to Cart