COMH4211AB70 Combi-Blocks, Inc.

Benzyl 2-amino-3-hydroxy-2-methylpropanoate

  • MDL number MFCD24644776
  • Cas Number : 160741-79-7
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1300

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1300
Availability details for
Quantity
  • Availability
  • Add to Cart