COMH4211AB76 Combi-Blocks, Inc.

5-Bromo-n-cyclobutylpyridin-2-amine

  • MDL number MFCD16747642
  • Cas Number : 1289058-28-1
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1304
Availability details for
Quantity
  • Availability
  • Add to Cart