COMH4211AB7A Combi-Blocks, Inc.

6-Chloro-2-(4-methoxy-benzyloxy)-3-nitro-pyridine

  • MDL number MFCD13656804
  • Cas Number : 1097080-93-7
  • Molecular Weight : 294.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1307
Availability details for
Quantity
  • Availability
  • Add to Cart