COMH4211AB82 Combi-Blocks, Inc.

4-(4-Bromo-3-methyl-phenylamino)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD12031011
  • Cas Number : 1134335-19-5
  • Molecular Weight : 369.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1317

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1317
Availability details for
Quantity
  • Availability
  • Add to Cart