COMH4211AB84 Combi-Blocks, Inc.

6-Bromo-n-propylpyridine-3-carboxamide

  • MDL number MFCD21781796
  • Cas Number : 1515673-74-1
  • Molecular Weight : 243.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1318

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1318
Availability details for
Quantity
  • Availability
  • Add to Cart