COMH4211AB86 Combi-Blocks, Inc.

2-Allyl-3-hydroxy-4-methoxybenzaldehyde

  • MDL number MFCD00266470
  • Cas Number : 16273-13-5
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1325
Availability details for
Quantity
  • Availability
  • Add to Cart