COMH4211AB88 Combi-Blocks, Inc.

2-(4-Trifluoromethoxy-phenoxy)-phenylamine

  • MDL number MFCD26687653
  • Cas Number : 150361-01-6
  • Molecular Weight : 269.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1327

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1327
Availability details for
Quantity
  • Availability
  • Add to Cart