COMH4211AB8C Combi-Blocks, Inc.

4-Methoxy-2-(2-methoxyethoxy)benzoic acid

  • MDL number MFCD14649660
  • Cas Number : 87359-71-5
  • Molecular Weight : 226.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1329
Availability details for
Quantity
  • Availability
  • Add to Cart