COMH4211AB8E Combi-Blocks, Inc.

4-(1H-Pyrazol-1-yl)cyclohexan-1-one

  • MDL number MFCD28363850
  • Cas Number : 1425971-78-3
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1H-pyrazol-1-yl)cyclohexan-1-one
Inchi Code 1S/C9H12N2O/c12-9-4-2-8(3-5-9)11-7-1-6-10-11/h1,6-8H,2-5H2
Inchi Key ICBCZKSSFUDQNY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1331

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1331
Availability details for
Quantity
  • Availability
  • Add to Cart