COMH4211AB90 Combi-Blocks, Inc.

1-(4-Bromo-2-fluoro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561524
  • Cas Number : 1596023-38-9
  • Molecular Weight : 271.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-1333
Availability details for
Quantity
  • Availability
  • Add to Cart